Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720543.3 | XP_006720606.1 | 17 | 16129 | ||
XM_006720544.3 | XP_006720607.1 | 16 | 16105 | |||
XM_017022240.1 | XP_016877729.1 | 18 | 16000 | |||
XM_017022241.1 | XP_016877730.1 | 17 | 15485 | |||
XM_017022243.1 | XP_016877732.1 | 17 | 15481 | |||
XM_017022244.1 | XP_016877733.1 | 17 | 15383 | |||
XM_017022245.1 | XP_016877734.1 | 18 | 15438 | |||
XM_017022246.1 | XP_016877735.1 | 17 | 15539 | |||
XM_017022251.1 | XP_016877740.1 | 10 | 14500 | |||
XM_017022252.1 | XP_016877741.1 | 9 | 14476 | |||
XM_017022253.1 | XP_016877742.1 | 18 | 16171 | |||
XM_017022254.1 | XP_016877743.1 | 18 | 16204 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL109696.1 | - | 2 | 1638 | Chr. 15 - 87863523 - 87865601 [-] on Build GRCh38 |