Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053055.4 | NP_444283.2 | 6 | 4182 | ||
GenBank mRNA | AK130716.1 | - | 1 | 1461 |