Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011525843.1 | XP_011524145.1 | 29 | 4887 | ||
GenBank mRNA | AK310385.1 | - | 26 | 3435 | ||
S81335.1 | - | 1 | 67 |