Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006714749.3 | XP_006714812.1 | 12 | 8414 | ||
XM_006714750.3 | XP_006714813.1 | 12 | 8353 | |||
XM_017008922.1 | XP_016864411.1 | 12 | 8700 | |||
XM_017008923.1 | XP_016864412.1 | 12 | 8669 | |||
GenBank mRNA | AK128316.1 | - | 1 | 4191 |