Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01422 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647841.1 | - | 3 | 4347 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000434868.1 | - | 3 | 4345 | Chr. 22 - 26858634 - 26865786 [+] on Build GRCh38 |