Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017016627.1 | XP_016872116.1 | 1 | 79 | ||
XM_017016628.1 | XP_016872117.1 | 1 | 79 | |||
XM_017016629.1 | XP_016872118.1 | 1 | 79 | |||
GenBank mRNA | BC014263.2 | - | 1 | 799 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD677920.1 | - | 1 | 397 | Chr. 10 - 68901733 - 68902213 [+] on Build GRCh38 |