Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC014263.2 | - | 1 | 388 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA772441.1 | - | 1 | 81 | Chr. 10 - 68901642 - 68902135 [+] on Build GRCh38 |