Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100128727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124397.1 | - | 1 | 1307 |
Gene Symbol: DGKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516684.2 | XP_011514986.2 | 27 | 15121 | ||
XM_017012780.1 | XP_016868269.1 | 34 | 7938 | |||
XM_017012781.1 | XP_016868270.1 | 34 | 7922 | |||
XM_017012782.1 | XP_016868271.1 | 33 | 7823 | |||
XM_017012783.1 | XP_016868272.1 | 33 | 7814 | |||
XM_017012784.1 | XP_016868273.1 | 32 | 7748 | |||
XM_017012785.1 | XP_016868274.1 | 32 | 8125 | |||
XM_017012786.1 | XP_016868275.1 | 34 | 7625 | |||
XM_017012787.1 | XP_016868276.1 | 33 | 7490 | |||
XM_017012788.1 | XP_016868277.1 | 34 | 7608 | |||
XM_017012789.1 | XP_016868278.1 | 20 | 6334 |