Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100128727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124397.1 | - | 1 | 2094 |
Gene Symbol: DGKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516684.2 | XP_011514986.2 | 27 | 15908 | ||
XM_017012780.1 | XP_016868269.1 | 34 | 8725 | |||
XM_017012781.1 | XP_016868270.1 | 34 | 8709 | |||
XM_017012782.1 | XP_016868271.1 | 33 | 8610 | |||
XM_017012783.1 | XP_016868272.1 | 33 | 8601 | |||
XM_017012784.1 | XP_016868273.1 | 32 | 8535 | |||
XM_017012785.1 | XP_016868274.1 | 32 | 8912 | |||
XM_017012786.1 | XP_016868275.1 | 34 | 8412 | |||
XM_017012787.1 | XP_016868276.1 | 33 | 8277 | |||
XM_017012788.1 | XP_016868277.1 | 34 | 8395 | |||
XM_017012789.1 | XP_016868278.1 | 20 | 7121 |