Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015134.3 | NP_055949.2 | 23 | 10516 | ||
NM_201274.3 | NP_958431.2 | 24 | 10579 | |||
XM_005256563.4 | XP_005256620.1 | 24 | 14628 | |||
XM_005256564.4 | XP_005256621.3 | 22 | 10707 | |||
XM_011523761.2 | XP_011522063.1 | 24 | 14999 | |||
XM_011523762.2 | XP_011522064.1 | 25 | 15045 | |||
XM_011523763.2 | XP_011522065.1 | 23 | 14868 | |||
XM_011523764.2 | XP_011522066.1 | 23 | 14862 | |||
XM_011523765.2 | XP_011522067.1 | 24 | 14925 | |||
XM_011523766.2 | XP_011522068.2 | 23 | 11175 | |||
XM_011523767.2 | XP_011522069.2 | 24 | 11238 | |||
XM_017024393.1 | XP_016879882.1 | 23 | 11118 | |||
GenBank mRNA | AK026035.1 | - | 1 | 972 | ||
AK091337.1 | - | 1 | 1819 |