Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TULP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535946.1 | XP_011534248.1 | 2 | 219 | ||
XM_017011063.1 | XP_016866552.1 | 3 | 368 | |||
XM_017011064.1 | XP_016866553.1 | 4 | 533 | |||
XM_017011065.1 | XP_016866554.1 | 3 | 286 | |||
XM_017011066.1 | XP_016866555.1 | 2 | 176 | |||
XM_017011067.1 | XP_016866556.1 | 2 | 269 | |||
XM_017011068.1 | XP_016866557.1 | 2 | 235 | |||
XM_017011069.1 | XP_016866558.1 | 3 | 371 | |||
XM_017011070.1 | XP_016866559.1 | 2 | 219 | |||
XM_017011071.1 | XP_016866560.1 | 3 | 371 | |||
GenBank mRNA | AF087973.1 | - | 520 |