Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF118093.1 | - | 1 | 992 | ||
AK126941.1 | - | 1 | 1770 | |||
BX537660.1 | - | 1 | 112 |