Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF086561.1 | - | 1 | 604 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AW750026.1 | - | 1 | 383 | Chr. 11 - 108147843 - 108148271 [+] on Build GRCh38 |