Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102450.2 | NP_001095920.1 | 7 | 4857 | ||
NM_033345.3 | NP_203131.1 | 6 | 4728 | |||
XM_011510089.2 | XP_011508391.1 | 7 | 5768 | |||
XM_011510090.2 | XP_011508392.1 | 10 | 5598 | |||
XM_017002631.1 | XP_016858120.1 | 10 | 5218 | |||
XM_017002632.1 | XP_016858121.1 | 8 | 5049 | |||
XM_017002633.1 | XP_016858122.1 | 7 | 5247 | |||
XM_017002634.1 | XP_016858123.1 | 7 | 4926 | |||
XM_017002635.1 | XP_016858124.1 | 8 | 5373 | |||
GenBank mRNA | AK057753.1 | - | 560 |