Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105374181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130735.1 | - | 1 | 1422 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA675109.1 | - | 1 | 155 | Chr. 3 - 158871162 - 158871745 [+] on Build GRCh38 |