Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUD6B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110439.1 | 2 | 1621 | |||
GenBank mRNA | BC030623.1 | - | 2 | 1626 | ||
BC105990.1 | - | 2 | 1528 |