Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003818.3 | NP_003809.1 | 13 | 4004 | ||
XM_006723660.1 | XP_006723723.1 | 13 | 4015 | |||
XM_011529390.1 | XP_011527692.1 | 14 | 4327 | |||
GenBank mRNA | AK130193.1 | - | 1 | 888 |