Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005246966.1 | XP_005247023.1 | 14 | 4903 | ||
XM_005246967.1 | XP_005247024.1 | 14 | 4950 | |||
GenBank mRNA | AK124958.1 | - | 1 | 503 |