Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NREP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021624.1 | - | 1 | 733 | ||
AK310585.1 | - | 2 | 977 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM876554.1 | - | 1 | 151 | Chr. 5 - 111734193 - 111734736 [-] on Build GRCh38 |