Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020980.1 | XP_016876469.1 | 4 | 2125 | ||
XM_017020981.1 | XP_016876470.1 | 3 | 1649 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK094455.1 | - | 1258 | Chr. - - [-] on Build |