Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100131878 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125681.1 | - | 1 | 335 |
Gene Symbol: NRDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021394.1 | XP_016876883.1 | 15 | 3619 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB003176.1 | - | 1 | 335 | Chr. 14 - 90274997 - 90275541 [-] on Build GRCh38 |