Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR3C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204265.1 | NP_001191194.1 | 7 | 3048 | ||
GenBank mRNA | U25029.1 | - | 1 | 533 |