Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR3C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204265.1 | NP_001191194.1 | 7 | 3424 | ||
GenBank mRNA | BF244382.1 | - | 1 | 336 | ||
U25029.1 | - | 1 | 909 |