Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005246467.3 | XP_005246524.1 | 15 | 1961 | ||
GenBank mRNA | BX641148.1 | - | 1 | 287 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB134028.1 | - | 1 | 176 | Chr. 2 - 190932528 - 190933067 [+] on Build GRCh38 |