Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256310.1 | NP_001243239.1 | 15 | 4030 | ||
XM_005246467.3 | XP_005246524.1 | 15 | 4089 | |||
GenBank mRNA | AF097494.1 | - | 1 | 1420 | ||
AF158555.1 | - | 16 | 4011 | |||
BX641148.1 | - | 1 | 2415 | |||
CR749593.1 | - | 8 | 4489 |