Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256310.1 | NP_001243239.1 | 15 | 1962 | ||
XM_005246467.3 | XP_005246524.1 | 15 | 2021 | |||
GenBank mRNA | AF097492.1 | - | 15 | 1596 | ||
AF158555.1 | - | 16 | 1942 | |||
AK001220.1 | - | 12 | 1034 | |||
BX641148.1 | - | 1 | 347 | |||
CR749593.1 | - | 8 | 2421 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB134028.1 | - | 1 | 236 | Chr. 2 - 190932528 - 190933067 [+] on Build GRCh38 |