Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005267730.3 | XP_005267787.1 | 8 | 3379 | ||
GenBank mRNA | AK057060.1 | - | 1 | 772 |