Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318067.1 | NP_001304996.1 | 13 | 4957 | ||
NM_001318068.1 | NP_001304997.1 | 13 | 5052 | |||
NM_001318069.1 | NP_001304998.1 | 14 | 5522 | |||
NM_002753.4 | NP_002744.1 | 14 | 5063 | |||
NM_138980.3 | NP_620446.1 | 14 | 5549 | |||
NM_138982.3 | NP_620448.1 | 14 | 5522 | |||
XM_005263131.2 | XP_005263188.1 | 14 | 5416 | |||
GenBank mRNA | AF052141.1 | - | 1 | 88 |