Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038227.1 | 4 | 531 | |||
NR_038228.1 | 4 | 518 | ||||
GenBank mRNA | AK055961.1 | - | 1 | 14 | ||
BC004962.1 | - | 4 | 531 | |||
BX109355.1 | - | 3 | 308 | |||
DA306887.1 | - | 4 | 442 | |||
DA682910.1 | - | 1 | 14 |