Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046251.1 | 2 | 882 | |||
NR_046252.1 | 2 | 872 | ||||
NR_046253.1 | 2 | 849 | ||||
NR_046254.1 | 2 | 845 | ||||
NR_046255.1 | 2 | 608 | ||||
GenBank mRNA | AK055924.1 | - | 2 | 850 | ||
AK096279.1 | - | 2 | 865 | |||
BC008609.1 | - | 2 | 551 | |||
BC039246.1 | - | 2 | 832 | |||
BC056913.1 | - | 2 | 608 | |||
BC092452.1 | - | 2 | 613 |