Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046251.1 | 2 | 3007 | |||
NR_046252.1 | 2 | 2997 | ||||
NR_046253.1 | 2 | 2974 | ||||
NR_046254.1 | 2 | 2970 | ||||
NR_046255.1 | 2 | 2733 | ||||
GenBank mRNA | BC039246.1 | - | 2 | 2957 | ||
BC042881.1 | - | 1 | 127 |