Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046251.1 | 2 | 2844 | |||
NR_046252.1 | 2 | 2834 | ||||
NR_046253.1 | 2 | 2811 | ||||
NR_046254.1 | 2 | 2807 | ||||
NR_046255.1 | 2 | 2570 | ||||
GenBank mRNA | BC039246.1 | - | 2 | 2794 |