Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK027156.1 | - | 1 | 1810 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA319421.1 | - | 1 | 29 | Chr. 11 - 117216848 - 117217186 [-] on Build GRCh38 |