Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1RL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282408.1 | NP_001269337.1 | 7 | 2739 | ||
NM_003856.3 | NP_003847.2 | 8 | 3059 | |||
NR_104167.1 | 9 | 3229 | ||||
XM_011512151.1 | XP_011510453.1 | 8 | 3066 | |||
GenBank mRNA | AL117622.1 | - | 1 | 656 |