Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC036550.1 | - | 1 | 2413 | ||
BC047239.1 | - | 1 | 2403 | |||
BC070105.1 | - | 1 | 1732 | |||
BC098409.1 | - | 1 | 1732 |