Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036519.1 | 2 | 529 | |||
XM_006722293.2 | XP_006722356.1 | 1 | 472 | |||
XM_011523606.2 | XP_011521908.1 | 1 | 472 | |||
XM_017025072.1 | XP_016880561.1 | 1 | 472 | |||
XM_017025073.1 | XP_016880562.1 | 1 | 472 | |||
XM_017025075.1 | XP_016880564.1 | 1 | 472 | |||
GenBank mRNA | AK090484.1 | - | 1 | 252 | ||
BX451530.2 | - | 1 | 91 | |||
DA436899.1 | - | 2 | 466 | |||
DA453235.1 | - | 2 | 466 |