Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF086543.1 | - | 1 | 197 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | W96291.1 | - | 1 | 197 | Chr. 11 - 35212550 - 35213708 [+] on Build GRCh38 |