Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242644.1 | NP_001229573.1 | 7 | 1407 | ||
NM_002865.2 | NP_002856.1 | 8 | 1479 | |||
GenBank mRNA | AF070629.1 | - | 4 | 900 | ||
AL137321.1 | - | 1 | 215 |