Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: API5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006718359.3 | XP_006718422.1 | 14 | 4087 | ||
XM_017018464.1 | XP_016873953.1 | 15 | 4144 | |||
GenBank mRNA | AK021649.1 | - | 1 | 1996 | ||
BC015898.1 | - | 1 | 50 |