Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199875.1 | NP_001186804.1 | 3 | 565 | ||
GenBank mRNA | AK307547.1 | - | 3 | 211 |