Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011541637.1 | XP_011539939.1 | 2 | 353 | ||
XM_011541640.1 | XP_011539942.1 | 2 | 353 | |||
XM_011541643.1 | XP_011539945.1 | 2 | 353 | |||
XM_017001544.1 | XP_016857033.1 | 1 | 322 | |||
XM_017001545.1 | XP_016857034.1 | 2 | 204 | |||
XM_017001548.1 | XP_016857037.1 | 2 | 353 | |||
XM_017001552.1 | XP_016857041.1 | 2 | 353 | |||
XM_017001553.1 | XP_016857042.1 | 2 | 212 | |||
XM_017001556.1 | XP_016857045.1 | 1 | 801 | |||
GenBank mRNA | AK056029.1 | - | 1 | 806 | ||
BC018109.1 | - | 2 | 233 | 3914920 |