Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012105.4 | NP_036237.2 | 9 | 6509 | ||
NM_138991.2 | NP_620476.1 | 8 | 6359 | |||
NM_138992.2 | NP_620477.1 | 8 | 6340 | |||
GenBank mRNA | AK023647.1 | - | 1 | 1015 | ||
BC020582.1 | - | 1 | 891 |