Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303451.1 | NP_001290380.1 | 4 | 6354 | ||
NM_024731.3 | NP_079007.2 | 5 | 6543 | |||
XM_005256149.2 | XP_005256206.1 | 5 | 6757 | |||
XM_017023674.1 | XP_016879163.1 | 5 | 6505 | |||
XM_017023675.1 | XP_016879164.1 | 5 | 6525 | |||
GenBank mRNA | AK056197.1 | - | 1 | 1537 | ||
AK056428.1 | - | 1 | 3341 | |||
AK074056.1 | - | 2 | 5470 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK057887.1 | - | 1377 | Chr. - - [-] on Build |