Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014171.5 | NP_054890.1 | 5 | 3575 | ||
GenBank mRNA | AK023264.1 | - | 1 | 925 |