Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011519011.2 | XP_011517313.1 | 28 | 6599 | ||
XM_011519012.2 | XP_011517314.1 | 28 | 6547 | |||
XM_011519013.2 | XP_011517315.1 | 28 | 7216 | |||
XM_011519014.2 | XP_011517316.1 | 28 | 6818 | |||
XM_011519015.1 | XP_011517317.1 | 29 | 5043 | |||
XM_011519016.1 | XP_011517318.1 | 27 | 4905 | |||
GenBank mRNA | AK094169.1 | - | 1 | 2067 |