Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AJAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011541786.2 | XP_011540088.1 | 7 | 6864 | ||
XM_011541787.2 | XP_011540089.1 | 6 | 6532 | |||
GenBank mRNA | CR936609.1 | - | 1 | 485 |