Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC038209.2 | - | 2 | 177 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI856750.1 | - | 3 | 195 | Chr. 7 - 20330389 - 20353091 [+] on Build GRCh38 |