Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACRGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513786.2 | XP_011512088.1 | 11 | 4554 | ||
XM_011513805.2 | XP_011512107.1 | 8 | 4095 | |||
XM_017007747.1 | XP_016863236.1 | 10 | 4309 | |||
XM_017007748.1 | XP_016863237.1 | 10 | 4260 | |||
XM_017007749.1 | XP_016863238.1 | 9 | 4175 | |||
XM_017007751.1 | XP_016863240.1 | 9 | 4377 | |||
XM_017007752.1 | XP_016863241.1 | 8 | 4224 | |||
XM_017007753.1 | XP_016863242.1 | 8 | 4094 | |||
XM_017007754.1 | XP_016863243.1 | 6 | 4151 | |||
XM_017007755.1 | XP_016863244.1 | 6 | 3968 | |||
XM_017007756.1 | XP_016863245.1 | 6 | 3961 | |||
XM_017007757.1 | XP_016863246.1 | 6 | 3897 | |||
XM_017007758.1 | XP_016863247.1 | 5 | 3846 | |||
XM_017007759.1 | XP_016863248.1 | 5 | 3794 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK125392.1 | - | 1 | 790 | Chr. 4 - 20729805 - 20732554 [+] on Build GRCh38 |