Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001843.3 | NP_001834.2 | 24 | 4302 | ||
NM_175038.2 | NP_778203.1 | 22 | 4118 | |||
XM_005268651.2 | XP_005268708.1 | 24 | 4665 | |||
XM_006719241.2 | XP_006719304.1 | 24 | 4289 | |||
XM_011537926.2 | XP_011536228.1 | 24 | 4254 | |||
XM_011537927.2 | XP_011536229.1 | 24 | 4311 | |||
GenBank mRNA | BX648591.1 | - | 1 | 1016 |