Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178040.1 | NP_001171511.1 | 10 | 3458 | ||
NM_004177.4 | NP_004168.1 | 11 | 3572 | |||
XM_005274200.3 | XP_005274257.1 | 10 | 3143 | |||
GenBank mRNA | AK055468.1 | - | 1 | 251 |